Learn More
Apexbio Technology LLC Isovaleryl-L-carnitine-d9-HCl
SDP
Catalog No. 5000570305
Encompass_Preferred
Isovaleryl-L-carnitine-d9 HCl (CAS No 1334532-23-8) is a deuterated derivative of L-carnitine primarily used in biomedical research to study metabolic pathways and cellular energy dynamics Originating from isotope labeling techniques this compound typically targets mitochondrial fatty acid metabolism facilitating the transport of fatty acids into mitochondria for -oxidation Its mechanism of action involves interaction with the carnitine shuttle system influencing lipid metabolism and energy production In vitro activity is generally observed in the nanomolar to low micromolar range Common applications include investigations using cell models such as hepatocytes or skeletal muscle cells and in vivo animal studies to understand metabolic disorders Furthermore it is utilized in drug screening to assess the impact on fatty acid oxidation pathways Experimental concentrations typically depend on the specific research design ensuring flexibility in various experimental setups
Catalog No. 50-005-70305 Supplier Apexbio Technology LLC Supplier No. SL10235
Isovaleryl-L-carnitine-d9 HCl (CAS No 1334532-23-8) is a deuterated derivative of L-carnitine primarily used in biomedical research to study metabolic pathways and cellular energy dynamics Originating from isotope labeling techniques this compound typically targets mitochondrial fatty acid metabolism facilitating the transport of fatty acids into mitochondria for -oxidation Its mechanism of action involves interaction with the carnitine shuttle system influencing lipid metabolism and energy production In vitro activity is generally observed in the nanomolar to low micromolar range Common applications include investigations using cell models such as hepatocytes or skeletal muscle cells and in vivo animal studies to understand metabolic disorders Furthermore it is utilized in drug screening to assess the impact on fatty acid oxidation pathways Experimental concentrations typically depend on the specific research design ensuring flexibility in various experimental setups