Learn More
Medchemexpress LLC Isovalerylcarnitine chloride | 139144-12-0 | 99.5% | C12H24ClNO4 | 100 MG
SDP
Catalog No. 5000356060
Encompass_Preferred
Isovalerylcarnitine chloride is a leucine metabolite that specifically activates human neutrophil calpain. Elevated circulating levels of this compound are inversely associated with a reduced risk of lung cancer. It is for research use only.
  • Metabolite of leucine
  • Specifically activates human neutrophil calpain
  • Inversely associated with a reduced risk of lung cancer
  • Available as a white to off-white solid
Catalog No. 50-003-56060 Supplier Medchemexpress LLC Supplier No. HY145542100MG
Isovalerylcarnitine chloride is a leucine metabolite that specifically activates human neutrophil calpain. Elevated circulating levels of this compound are inversely associated with a reduced risk of lung cancer. It is for research use only.
  • Metabolite of leucine
  • Specifically activates human neutrophil calpain
  • Inversely associated with a reduced risk of lung cancer
  • Available as a white to off-white solid