Learn More
Medchemexpress LLC Isovalerylcarnitine chloride | 139144-12-0 | 99.5% | C12H24ClNO4 | 10MG
SDP
Catalog No. 5000260210
Encompass_Preferred
Isovalerylcarnitine chloride is a research-grade biochemical reagent (CAS 139144-12-0) derived from L-leucine catabolism that functions as a potent activator of Ca2+-dependent calpain in human neutrophils. Supplied as a high-purity solid for in vitro biochemical and cell-based assays; intended for research use only.
  • High purity (99.48%) suitable for analytical and biological studies.
  • Activates Ca2+-dependent calpain in human neutrophils for pathway and mechanism studies.
  • Soluble in DMSO for convenient stock solution preparation.
  • Available in small quantities appropriate for screening and assay development.
  • Provided with product-specific data and handling recommendations for laboratory use.
Catalog No. 50-002-60210 Supplier Medchemexpress LLC Supplier No. HY14554210MG
Isovalerylcarnitine chloride is a research-grade biochemical reagent (CAS 139144-12-0) derived from L-leucine catabolism that functions as a potent activator of Ca2+-dependent calpain in human neutrophils. Supplied as a high-purity solid for in vitro biochemical and cell-based assays; intended for research use only.
  • High purity (99.48%) suitable for analytical and biological studies.
  • Activates Ca2+-dependent calpain in human neutrophils for pathway and mechanism studies.
  • Soluble in DMSO for convenient stock solution preparation.
  • Available in small quantities appropriate for screening and assay development.
  • Provided with product-specific data and handling recommendations for laboratory use.