Learn More
Medchemexpress LLC Isovalerylcarnitine chloride | 139144-12-0 | MFCD09263554 | 99.5% | 281.78 g·mol-1 | C12H24ClNO4 | 5 MG
SDP
Catalog No. 5000221341
Encompass_Preferred
Isovalerylcarnitine chloride is the chloride salt of isovalerylcarnitine, a metabolic product of L-leucine that acts as a potent activator of Ca2+-dependent protease (calpain). Supplied as a research-grade analytical standard, it is intended for in vitro and analytical use and is accompanied by a datasheet, COA, and SDS.
Catalog No. 50-002-21341 Supplier Medchemexpress LLC Supplier No. HY1455425MG
Isovalerylcarnitine chloride is the chloride salt of isovalerylcarnitine, a metabolic product of L-leucine that acts as a potent activator of Ca2+-dependent protease (calpain). Supplied as a research-grade analytical standard, it is intended for in vitro and analytical use and is accompanied by a datasheet, COA, and SDS.