Learn More
Cayman Chemical Isovaleryl-L-carnitine (chloride), Purity: ≥95%, CAS: 139144-12-0, 100 mg
SDP
Catalog No. 502933481
Encompass_Preferred
A naturally occurring acylcarnitine; formed via metabolic conversion of L-leucine; increases survival and decreases apoptosis in hepatocyte growth factor-deprived murine C2.8 hepatocytes at 1 mM; inhibits amino acid deprivation-induced proteolysis and autophagy in isolated perfused rat liver at 77 and 100 μM, respectively
Catalog No. 50-293-3481 Supplier Cayman Chemical Supplier No. 26555100 MG
A naturally occurring acylcarnitine; formed via metabolic conversion of L-leucine; increases survival and decreases apoptosis in hepatocyte growth factor-deprived murine C2.8 hepatocytes at 1 mM; inhibits amino acid deprivation-induced proteolysis and autophagy in isolated perfused rat liver at 77 and 100 μM, respectively