Learn More
Medchemexpress LLC Isovanillic acid | 645-08-9 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN686410MM/1ML
Encompass_Preferred
Isovanillic acid (3-Hydroxy-4-methoxybenzoic acid) is a phenolic acid isolated from Scrophularia ningpoensis. It exhibits anti-thrombotic activity by inhibiting fibrin and blood clot formation, prolonging partial thromboplastin time, and increasing platelet aggregation closure time. This compound also shows potential in research for insulin resistance and type 2 diabetes by increasing 2-[14C(U)]-deoxy-D-glucose uptake in LHCN-M2 cells.
  • Phenolic acid isolated from Scrophularia ningpoensis.
  • Exhibits anti-thrombotic activity.
  • Potential for research in insulin resistance and type 2 diabetes.
  • Slight uncompetitive inhibitory activity against thrombin.
  • Inhibits fibrin clot and blood clot formation.
  • Prolongs partial thromboplastin time.
  • Increases closure time of platelet aggregation.
Catalog No. 50-004-20996