Learn More
Medchemexpress LLC Isovanillic acid | 645-08-9 | 5 MG
SDP
Catalog No. 5000417204
Encompass_Preferred
Isovanillic acid (3-Hydroxy-4-methoxybenzoic acid) is a phenolic acid isolated from *Scrophularia ningpoensis*. It exhibits anti-thrombotic activity and is involved in various biological processes.
  • Increases 2-[14C(U)]-deoxy-D-glucose (2-DG) uptake in LHCN-M2 cells, which has potential in research of insulin resistance and type 2 diabetes
  • Exhibits uncompetitive inhibitory activity against thrombin
  • Inhibits the formation of fibrin clot and blood clot
  • Prolongs partial thromboplastin time
  • Increases platelet aggregation closure time, showing antithrombotic, anticoagulant, and antiplatelet activities
Catalog No. 50-004-17204 Supplier Medchemexpress LLC Supplier No. HYN68645MG
Isovanillic acid (3-Hydroxy-4-methoxybenzoic acid) is a phenolic acid isolated from *Scrophularia ningpoensis*. It exhibits anti-thrombotic activity and is involved in various biological processes.
  • Increases 2-[14C(U)]-deoxy-D-glucose (2-DG) uptake in LHCN-M2 cells, which has potential in research of insulin resistance and type 2 diabetes
  • Exhibits uncompetitive inhibitory activity against thrombin
  • Inhibits the formation of fibrin clot and blood clot
  • Prolongs partial thromboplastin time
  • Increases platelet aggregation closure time, showing antithrombotic, anticoagulant, and antiplatelet activities