Learn More
Enzo Life Sciences Isovelleral (1mg). CAS: 37841-91-1
SDP
Supplier:  Enzo Life Sciences ALX550356M001
Encompass_Preferred
Antagonist with different binding profiles towards the vanilloid receptor 1 (TRPV1). Activator of TRPA1 Purity: ≥99%. Solubility: Soluble in organic solvents. Long Term Storage: -20°C.
Catalog No. 50-200-9540