Learn More
Cayman Chemical IsoverbascosIde 50mg
SDP
Catalog No. 502939024
Encompass_Preferred
A phenylethanoid glycoside with diverse biological activities; scavenges DPPH radicals in a cell-free assay and inhibits hydrogen peroxide-induced lipid peroxidation in V79-4 cells at 10 μg/ml; induces apoptosis and production of ROS in, and reduces viability of, OVCAR-3 cells from 7.5-30 μM; inhibits tumor growth in an OVCAR-3 mouse xenograft model at 30 mg/kg; decreases brain amyloid deposition and increases exploratory behavior in Aβ42-infused rats at 2.5 and 5 mg/kg; decreases xylene-induced EAr edema in mice and increases survival in a mouse model of LPS-induced endotoxic shock
Catalog No. 50-293-9024 Supplier Cayman Chemical Supplier No. 2921650 MG
A phenylethanoid glycoside with diverse biological activities; scavenges DPPH radicals in a cell-free assay and inhibits hydrogen peroxide-induced lipid peroxidation in V79-4 cells at 10 μg/ml; induces apoptosis and production of ROS in, and reduces viability of, OVCAR-3 cells from 7.5-30 μM; inhibits tumor growth in an OVCAR-3 mouse xenograft model at 30 mg/kg; decreases brain amyloid deposition and increases exploratory behavior in Aβ42-infused rats at 2.5 and 5 mg/kg; decreases xylene-induced EAr edema in mice and increases survival in a mouse model of LPS-induced endotoxic shock