Learn More
Medchemexpress LLC Isovitexin 2"-O-arabinoside | 53382-71-1 | 5 MG
SDP
Catalog No. 5000419810
Encompass_Preferred
Isovitexin 2''-O-arabinoside is a flavonoid found in plantlets of Avena sativa L. It increases alkaline phosphatase (ALP) activity and promotes BMP-2-induced osteoblast differentiation in C2C12 cells at concentrations from 0-10 μM over 3 days. It is for research use only.
  • Flavonoid found in Avena sativa L. plantlets
  • Increases alkaline phosphatase (ALP) activity
  • Promotes BMP-2-induced osteoblast differentiation
  • Targets phosphatase
  • Involved in metabolic enzyme/protease pathway
Catalog No. 50-004-19810 Supplier Medchemexpress LLC Supplier No. HYN51145MG
Isovitexin 2''-O-arabinoside is a flavonoid found in plantlets of Avena sativa L. It increases alkaline phosphatase (ALP) activity and promotes BMP-2-induced osteoblast differentiation in C2C12 cells at concentrations from 0-10 μM over 3 days. It is for research use only.
  • Flavonoid found in Avena sativa L. plantlets
  • Increases alkaline phosphatase (ALP) activity
  • Promotes BMP-2-induced osteoblast differentiation
  • Targets phosphatase
  • Involved in metabolic enzyme/protease pathway