Learn More
Medchemexpress LLC HY-N0773 5mg Medchemexpress, Isovitexin CAS:38953-85-4 Purity:>98%
SDP
Catalog No. 501933710
Encompass_Preferred
Medchemexpress, HY-N0773 5mg Isovitexin CAS:38953-85-4 Isovitexin is a flavonoid isolated from rice hulls of Oryza sativa, possesses anti-inflammatory and anti-oxidant activities; Isovitexin acts like a JNK1/2 inhibitor and inhibits the activation of NF-κB . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3710 Supplier Medchemexpress LLC Supplier No. HYN07735MG
Medchemexpress, HY-N0773 5mg Isovitexin CAS:38953-85-4 Isovitexin is a flavonoid isolated from rice hulls of Oryza sativa, possesses anti-inflammatory and anti-oxidant activities; Isovitexin acts like a JNK1/2 inhibitor and inhibits the activation of NF-κB . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.