Learn More
Medchemexpress LLC Isoxadifen-ethyl | 163520-33-0 | ≥97.0% | 1 ML
SDP
Catalog No. 5000339859
Encompass_Preferred
Isoxadifen-ethyl is a widely used herbicide safener in rice that can decrease the phytotoxicity caused by fenoxaprop-P-ethyl. It is for research use only.
  • Functions as a widely used herbicide safener.
  • Decreases phytotoxicity caused by fenoxaprop-P-ethyl.
  • Intended for research use only.
Catalog No. 50-003-39859 Supplier Medchemexpress LLC Supplier No. HY13036110MM/1ML
Isoxadifen-ethyl is a widely used herbicide safener in rice that can decrease the phytotoxicity caused by fenoxaprop-P-ethyl. It is for research use only.
  • Functions as a widely used herbicide safener.
  • Decreases phytotoxicity caused by fenoxaprop-P-ethyl.
  • Intended for research use only.