Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isoxadifen-ethyl | 163520-33-0 | ≥97.0% | 5 G
SDP

Catalog No. 5000338335
Encompass_Preferred

Isoxadifen-ethyl is a widely used compound that functions as an herbicide safener, particularly in rice. Its primary role is to mitigate the phytotoxicity that can arise from the application of fenoxaprop-P-ethyl, thereby protecting crops from damage. This compound is intended for research use only.

  • Used as an herbicide safener in rice.
  • Decreases phytotoxicity caused by fenoxaprop-P-ethyl.
  • Chemical Formula: C18H17NO3.
  • Molecular Weight: 295.34.
  • Appearance: Solid, white to light yellow.

Catalog No. 50-003-38335 Supplier Medchemexpress LLC Supplier No. HY1303615G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isoxadifen-ethyl is a widely used compound that functions as an herbicide safener, particularly in rice. Its primary role is to mitigate the phytotoxicity that can arise from the application of fenoxaprop-P-ethyl, thereby protecting crops from damage. This compound is intended for research use only.

  • Used as an herbicide safener in rice.
  • Decreases phytotoxicity caused by fenoxaprop-P-ethyl.
  • Chemical Formula: C18H17NO3.
  • Molecular Weight: 295.34.
  • Appearance: Solid, white to light yellow.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.