Learn More
Enzo Life Sciences Isoxanthohumol (1mg). CAS: 521-48-2
SDP
Supplier:  Enzo Life Sciences ALX350279M001
Encompass_Preferred
Racemic mixture of (+/-)-Isoxanthohumol. Prenylated flavonoid. Phytoestrogen. Induces apoptosis in mature adipocytes. Alternative name: (+/-)-Isoxanthohumol. Purity: ≥98% (1H-NMR). Solubility: Soluble in DMSO (50mg/ml) or methanol (50mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-8825