Learn More
AdipoGen Isoxathion
SDP
Supplier:  AdipoGen CDXI0049M500
Encompass_Preferred
Chemical. CAS 18854-01-8. Formula C13H16NO4PS. MW 313.3. Synthetic. Isoxazole organothiophosphate insecticide. Works as an acetylcholine esterase inhibitor. Compound can be used as analytical reference material.
Catalog No. 50-256-0638