Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isoxazol-3-amine | 1750-42-1 | 84.08 | 25 G
SDP

Catalog No. 5000448932
Encompass_Preferred

Isoxazol-3-amine is a biochemical reagent designed for life science research. It can be utilized as a biological material or an organic compound in various research applications. This product is intended for research use only and is not for sale to patients.

  • Purity of 98.9%
  • Molecular formula C3H4N2O
  • Molecular weight of 84.08
  • CAS number 1750-42-1
  • Appears as solid below 23°C, liquid above 23°C
  • Colorless to light yellow
  • Store at 4°C, protected from light
  • For in-solvent storage: -80°C for 6 months, or -20°C for 1 month, both protected from light
  • In vitro solubility in DMSO is 100 mg/mL (1189.34 mM) with ultrasonic and warming to 60°C

Catalog No. 50-004-48932 Supplier Medchemexpress LLC Supplier No. 25GHYW007329
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isoxazol-3-amine is a biochemical reagent designed for life science research. It can be utilized as a biological material or an organic compound in various research applications. This product is intended for research use only and is not for sale to patients.

  • Purity of 98.9%
  • Molecular formula C3H4N2O
  • Molecular weight of 84.08
  • CAS number 1750-42-1
  • Appears as solid below 23°C, liquid above 23°C
  • Colorless to light yellow
  • Store at 4°C, protected from light
  • For in-solvent storage: -80°C for 6 months, or -20°C for 1 month, both protected from light
  • In vitro solubility in DMSO is 100 mg/mL (1189.34 mM) with ultrasonic and warming to 60°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.