Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isoxazol-3-amine | 1750-42-1 | 84.08 | 5 G
SDP

Catalog No. 5000460277
Encompass_Preferred

Isoxazol-3-amine is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Appears as solid below 23°C and liquid above 23°C
  • Colorless to light yellow
  • Soluble in DMSO: 100 mg/mL (1189.34 mM) with ultrasonic and warming to 60°C
  • Shipping at room temperature in continental US; may vary elsewhere
  • Storage at 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light)

Catalog No. 50-004-60277 Supplier Medchemexpress LLC Supplier No. HYW0073295G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isoxazol-3-amine is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Appears as solid below 23°C and liquid above 23°C
  • Colorless to light yellow
  • Soluble in DMSO: 100 mg/mL (1189.34 mM) with ultrasonic and warming to 60°C
  • Shipping at room temperature in continental US; may vary elsewhere
  • Storage at 4°C, protect from light. In solvent: -80°C for 6 months; -20°C for 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.