Learn More
Medchemexpress LLC Isoxazole | 288-14-2 | 100.0% | 69.06 | 1 ML
SDP
Catalog No. 5000453950
Encompass_Preferred
Isoxazole is a five-membered heterocycle drug scaffold that has been used as a BET bromodomain inhibitor and can improve β-cell function in a diabetic mouse model. Its derivatives show broad biological activities, including antimicrobial, antibacterial, antifungal, antiviral, anticancer, anti-inflammatory, immunomodulatory, analgesic, anti-tuberculosis, and anti-diabetic effects.
  • Broad biological activities: Isoxazole and its derivatives exhibit a wide range of biological activities.
  • BET bromodomain inhibitor: It has been used as a BET bromodomain inhibitor.
  • Improved β-cell function: It can improve β-cell function in a diabetic mouse model.
  • HSP90 inhibitor: Bicyclic Isoxazole can act as an HSP90 inhibitor.
  • Selective multidrug resistance protein (MRP1) inhibitor: Tricyclic Isoxazole shows promise as a selective MRP1 inhibitor.
Catalog No. 50-004-53950 Supplier Medchemexpress LLC Supplier No. HYW01064910MM/1ML
Isoxazole is a five-membered heterocycle drug scaffold that has been used as a BET bromodomain inhibitor and can improve β-cell function in a diabetic mouse model. Its derivatives show broad biological activities, including antimicrobial, antibacterial, antifungal, antiviral, anticancer, anti-inflammatory, immunomodulatory, analgesic, anti-tuberculosis, and anti-diabetic effects.
  • Broad biological activities: Isoxazole and its derivatives exhibit a wide range of biological activities.
  • BET bromodomain inhibitor: It has been used as a BET bromodomain inhibitor.
  • Improved β-cell function: It can improve β-cell function in a diabetic mouse model.
  • HSP90 inhibitor: Bicyclic Isoxazole can act as an HSP90 inhibitor.
  • Selective multidrug resistance protein (MRP1) inhibitor: Tricyclic Isoxazole shows promise as a selective MRP1 inhibitor.