Learn More
Selleck Chemical LLC Isoxazole 9
SDP
Catalog No. 5000003890
Encompass_Preferred
Isoxazole 9 (Isx-9) is a synthetic promotor of adult neurogenesis by triggering neuronal differentiation of adult neural stem/precursor cells (NSPCs) Isoxazole 9 (Isx-9) activates multiple pathways including TGF- induced epithelial mesenchymal transition (EMT) signaling canonical and non-canonical Wnt signaling at different stages of cardiac differentiation
Catalog No. 50-000-03890 Supplier Selleck Chemical LLC Supplier No. S791425MG
Isoxazole 9 (Isx-9) is a synthetic promotor of adult neurogenesis by triggering neuronal differentiation of adult neural stem/precursor cells (NSPCs) Isoxazole 9 (Isx-9) activates multiple pathways including TGF- induced epithelial mesenchymal transition (EMT) signaling canonical and non-canonical Wnt signaling at different stages of cardiac differentiation