Learn More
Selleck Chemical LLC Isoxazole 9
SDP
Supplier:  Selleck Chemical LLC S7914100MG
Encompass_Preferred
Isoxazole 9 (Isx-9) is a synthetic promotor of adult neurogenesis by triggering neuronal differentiation of adult neural stem/precursor cells (NSPCs) Isoxazole 9 (Isx-9) activates multiple pathways including TGF- induced epithelial mesenchymal transition (EMT) signaling canonical and non-canonical Wnt signaling at different stages of cardiac differentiation
Catalog No. 50-000-01314