Learn More
Medchemexpress LLC Isoxazolium, 2-ethyl-5-(3-sulfophenyl)-, inner salt | 4156-16-5 | MFCD00012128 | 98.0% | 253.27 g/mol | C11H11NO4S | 250 MG
SDP
Catalog No. 5000282078
Encompass_Preferred
Woodward's Reagent K is an isoxazolium-derived biochemical reagent supplied for laboratory research use. It is provided with analytical documentation to support experimental reproducibility and safe handling. Suitable for derivatization and detection workflows, the reagent is offered in multiple pack sizes and grades to fit different experimental scales.
  • Isoxazolium-based reagent for derivatization reactions.
  • High purity suitable for analytical and research applications.
  • Available in multiple pack sizes for flexible use.
  • Includes COA and SDS for quality and safety documentation.
  • Compatible with common analytical techniques such as mass spectrometry.
Catalog No. 50-002-82078 Supplier Medchemexpress LLC Supplier No. HYW341151250MG
Woodward's Reagent K is an isoxazolium-derived biochemical reagent supplied for laboratory research use. It is provided with analytical documentation to support experimental reproducibility and safe handling. Suitable for derivatization and detection workflows, the reagent is offered in multiple pack sizes and grades to fit different experimental scales.
  • Isoxazolium-based reagent for derivatization reactions.
  • High purity suitable for analytical and research applications.
  • Available in multiple pack sizes for flexible use.
  • Includes COA and SDS for quality and safety documentation.
  • Compatible with common analytical techniques such as mass spectrometry.