Learn More
Medchemexpress LLC Isoxazolo-pyridinone 7e | 500164-74-9 | 99.7% | 390.43 | 50 MG
SDP
Catalog No. 5000343598
Encompass_Preferred
IP7e is a potent, brain-penetrant, and orally active Nurr1 activator. It is intended for research use only.
  • Activator of Nurr1 with an EC50 value of 3.9 nM.
  • Reduces incidence and severity of experimental autoimmune encephalomyelitis (EAE) in a murine model with preventive treatment.
  • Attenuates inflammation and neurodegeneration in spinal cords of EAE mice.
  • Acts through an NF-κB pathway-dependent process.
Catalog No. 50-003-43598 Supplier Medchemexpress LLC Supplier No. HY11027410MG
IP7e is a potent, brain-penetrant, and orally active Nurr1 activator. It is intended for research use only.
  • Activator of Nurr1 with an EC50 value of 3.9 nM.
  • Reduces incidence and severity of experimental autoimmune encephalomyelitis (EAE) in a murine model with preventive treatment.
  • Attenuates inflammation and neurodegeneration in spinal cords of EAE mice.
  • Acts through an NF-κB pathway-dependent process.