Learn More
Medchemexpress LLC Isoxicam | 34552-84-6 | 99.8% | 250 MG
SDP
Catalog No. 5000414121
Encompass_Preferred
Isoxicam is an orally active, long-acting, non-steroidal anti-inflammatory agent for the research of arthritis. It is a nonselective inhibitor of COX-1 and COX-2.
  • Orally active
  • Long-acting
  • Non-steroidal anti-inflammatory agent
  • Nonselective inhibitor of COX-1 and COX-2
  • Used for arthritis research
Catalog No. 50-004-14121 Supplier Medchemexpress LLC Supplier No. 250MGHYB1130
Isoxicam is an orally active, long-acting, non-steroidal anti-inflammatory agent for the research of arthritis. It is a nonselective inhibitor of COX-1 and COX-2.
  • Orally active
  • Long-acting
  • Non-steroidal anti-inflammatory agent
  • Nonselective inhibitor of COX-1 and COX-2
  • Used for arthritis research