Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoxicam, MilliporeSigma™ Supelco™

Catalog No. 111013986 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3986 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3986 Supplier MilliporeSigma Supelco Supplier No. I17621G
Add to Cart
Add to Cart
Isoxicam is a long acting anti-inflammatory agent belonging to the oxicam group, and helps in relieving the symptoms of degenerative joint disease and rheumatoid arthritis. Its mode of action involves the ability to inhibit prostaglandin synthesis by suppressing the formation of cyclooxygenase and subsequent prostaglandin. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 34552-84-6
Physical Form Neat
Linear Formula C14H13N3O5 S
MDL Number MFCD00079374
Type Analytical Standard
Quantity 1 g
Formula Weight 335.34
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.