Learn More
Cayman Chemical IsoxnthopterIn 100mg
SDP
Catalog No. 502967652
Encompass_Preferred
A natural intermediate in the pteridine pathway that can be produced naturally from 7-oxobiopterin or in vitro by the oxidation of pterin by xanthine oxidase
Catalog No. 50-296-7652 Supplier Cayman Chemical Supplier No. 17564100 MG
A natural intermediate in the pteridine pathway that can be produced naturally from 7-oxobiopterin or in vitro by the oxidation of pterin by xanthine oxidase