Learn More
Cayman Chemical IsoxnthopterIn 10mg
SDP
Catalog No. 502967638
Encompass_Preferred
A natural intermediate in the pteridine pathway that can be produced naturally from 7-oxobiopterin or in vitro by the oxidation of pterin by xanthine oxidase
Catalog No. 50-296-7638 Supplier Cayman Chemical Supplier No. 1756410 MG
A natural intermediate in the pteridine pathway that can be produced naturally from 7-oxobiopterin or in vitro by the oxidation of pterin by xanthine oxidase