Learn More
Cayman Chemical Isoxanthopterin, Purity: ≥90%, CAS: 529-69-1, 50 mg
SDP
Catalog No. 502967128
Encompass_Preferred
A natural intermediate in the pteridine pathway that can be produced naturally from 7-oxobiopterin or in vitro by the oxidation of pterin by xanthine oxidase
Catalog No. 50-296-7128 Supplier Cayman Chemical Supplier No. 1756450 MG
A natural intermediate in the pteridine pathway that can be produced naturally from 7-oxobiopterin or in vitro by the oxidation of pterin by xanthine oxidase