Learn More
Medchemexpress LLC Isoxsuprine-d6 (hydrochloride) | 2706004-35-3 | 99.9% | 500 UG
SDP
Catalog No. 5000412820
Encompass_Preferred
Isoxsuprine-d6 (hydrochloride) is the deuterium-labeled Isoxsuprine hydrochloride. Isoxsuprine hydrochloride is a beta-adrenergic receptor agonist with Kis of 13.65 μΜ and 3.48 μΜ for myometrial and placental beta-adrenergic receptors, respectively. It is also an NMDA receptor antagonist.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12820 Supplier Medchemexpress LLC Supplier No. HYB1270S500UG
Isoxsuprine-d6 (hydrochloride) is the deuterium-labeled Isoxsuprine hydrochloride. Isoxsuprine hydrochloride is a beta-adrenergic receptor agonist with Kis of 13.65 μΜ and 3.48 μΜ for myometrial and placental beta-adrenergic receptors, respectively. It is also an NMDA receptor antagonist.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.