Learn More
Apexbio Technology LLC Isoxsuprine (hydrochloride) 579-56-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000091440
Encompass_Preferred
Isoxsuprine hydrochloride (CAS 579-56-6) is a small molecule that modulates 2-adrenergic receptors a class of G protein-coupled receptors responsive to epinephrine and implicated in sympathetic nervous system signaling including smooth muscle relaxation and vasodilation Isoxsuprine functions as both an agonist and antagonist at these receptors In vitro Isoxsuprine has demonstrated preservation of neuronal viability when administered following hypoxic insult In vivo studies using stroke models show that post-ischemic administration of Isoxsuprine significantly reduces infarct volume These properties support its use in investigating neuroprotection cerebrovascular modulation and smooth muscle physiology
Catalog No. 50-000-91440 Supplier Apexbio Technology LLC Supplier No. C33475.1
Isoxsuprine hydrochloride (CAS 579-56-6) is a small molecule that modulates 2-adrenergic receptors a class of G protein-coupled receptors responsive to epinephrine and implicated in sympathetic nervous system signaling including smooth muscle relaxation and vasodilation Isoxsuprine functions as both an agonist and antagonist at these receptors In vitro Isoxsuprine has demonstrated preservation of neuronal viability when administered following hypoxic insult In vivo studies using stroke models show that post-ischemic administration of Isoxsuprine significantly reduces infarct volume These properties support its use in investigating neuroprotection cerebrovascular modulation and smooth muscle physiology