Learn More
Medchemexpress LLC Isoxsuprine hydrochloride | 579-56-6 | 99.95% | 1 ML
SDP
Catalog No. 5000412947
Encompass_Preferred
Isoxsuprine hydrochloride is a beta-adrenergic receptor agonist and an NMDA receptor antagonist. It inhibits circular chemorepellent induced defect (CCID) formation and 12(S)-HETE synthesis, and can reduce infarct volume.
  • Beta-adrenergic receptor agonist
  • NMDA receptor antagonist
  • Inhibits circular chemorepellent induced defect (CCID) formation
  • Inhibits 12(S)-HETE synthesis
  • Reduces infarct volume in vivo
Catalog No. 50-004-12947 Supplier Medchemexpress LLC Supplier No. HYB127010MM/1ML
Isoxsuprine hydrochloride is a beta-adrenergic receptor agonist and an NMDA receptor antagonist. It inhibits circular chemorepellent induced defect (CCID) formation and 12(S)-HETE synthesis, and can reduce infarct volume.
  • Beta-adrenergic receptor agonist
  • NMDA receptor antagonist
  • Inhibits circular chemorepellent induced defect (CCID) formation
  • Inhibits 12(S)-HETE synthesis
  • Reduces infarct volume in vivo