Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoxsuprine hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111020068 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0068 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0068 Supplier MilliporeSigma Supelco Supplier No. I08805G
Add to Cart
Add to Cart

Isoxsuprine hydrochloride is a drug, which is mainly used in the treatment of navicular syndrome in equines.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 579-56-6
Physical Form Neat
Linear Formula C18H23 NO3 · HCl
MDL Number MFCD00058069
Type Analytical Standard
Quantity 5 g
Formula Weight 337.84
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.