Learn More
Medchemexpress LLC Ispinesib | 336113-53-2 | 99.6% | C30H33ClN4O2 | 100 MG
SDP
Catalog No. 5000358146
Encompass_Preferred
Ispinesib is a specific inhibitor of kinesin spindle protein (KSP), exhibiting a Ki app of 1.7 nM.
  • Potent and highly specific inhibition of KSP.
  • Inhibits the growth of BT-474 and MDA-MB-468 cell lines.
  • Suppresses proliferation and induces apoptosis in prostate cancer cells.
  • Up-regulates genes for apoptosis and cell cycle arrest, while down-regulating genes for cell proliferation and survival.
  • Anti-proliferation and pro-apoptotic activities can be enhanced in combination with genistein.
  • Reduces tumor volume in mouse models bearing various breast cancer xenografts.
Catalog No. 50-003-58146 Supplier Medchemexpress LLC Supplier No. HY50759100MG
Ispinesib is a specific inhibitor of kinesin spindle protein (KSP), exhibiting a Ki app of 1.7 nM.
  • Potent and highly specific inhibition of KSP.
  • Inhibits the growth of BT-474 and MDA-MB-468 cell lines.
  • Suppresses proliferation and induces apoptosis in prostate cancer cells.
  • Up-regulates genes for apoptosis and cell cycle arrest, while down-regulating genes for cell proliferation and survival.
  • Anti-proliferation and pro-apoptotic activities can be enhanced in combination with genistein.
  • Reduces tumor volume in mouse models bearing various breast cancer xenografts.