Learn More
Medchemexpress LLC HY-50759 5mg , Ispinesib CAS:336113-53-2 Purity:>98%
SDP
Catalog No. 502259156
Encompass_Preferred
HY-50759 5mg Ispinesib CAS: 336113-53-2 Ispinesib is a specific inhibitor of kinesin spindle protein (KSP), with a Ki app of 1.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9156 Supplier Medchemexpress LLC Supplier No. HY507595MG
HY-50759 5mg Ispinesib CAS: 336113-53-2 Ispinesib is a specific inhibitor of kinesin spindle protein (KSP), with a Ki app of 1.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.