Learn More
Abcam Ispinesib (SB-715992), 10MG
Catalog No. 502682944
Encompass_Preferred
MW 517.1 Da, Purity >98%. A potent, selective and reversible allosteric inhibitor of KSP with IC₅₀ value of 0.5 nM. Often combines with chemotherapy drugs to tumor treatment.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2944 Supplier Abcam Supplier No. AB27352110MG
MW 517.1 Da, Purity >98%. A potent, selective and reversible allosteric inhibitor of KSP with IC₅₀ value of 0.5 nM. Often combines with chemotherapy drugs to tumor treatment.
The product is subject to the following: Abcam Restricted Use Statement