Learn More
LAB ALLEY Lab Alley Isopropyl Alcohol 91% Lab Grade
SDP
Supplier:  LAB ALLEY IPAL911GALCS
Encompass
Isopropyl Alcohol (IPA) at the 91% concentration is hydrophobic and evaporates quickly making this concentration ideal for use with products and environments that are sensitive to water and residues. Isopropyl alcohol is a colorless and flammable liquid. Lab grade chemicals are high quality but unlike the ACS/USP grade the exact impurities are unknown making these great for lab and research purposes but not pure enough for food or drug uses. This product contains 91% isopropyl alcohol 9% distilled water. The combination of alcohol and water makes this an ideal solution for cleaning and disinfecting. The presence of water makes this ideal for cleaning as it doesn’t evaporate as readily as a higher concentration. Isopropyl Alcohol is a colorless and flammable liquid.
Catalog No. NC3377782