Learn More
Medchemexpress LLC Isradipine | 75695-93-1 | 5 MG
SDP
Catalog No. 5000406124
Encompass_Preferred
Isradipine is an orally active, blood-brain barrier permeable L-type calcium channel blocker. It functions as a potent peripheral vasodilator and is a dihydropyridine calcium antagonist with selective effects on the heart and peripheral circulation. Isradipine is considered a potentially viable neuroprotective agent for Parkinson's disease.
  • Orally active
  • Blood-brain barrier permeable
  • L-type calcium channel blocker
  • Potent peripheral vasodilator
  • Dihydropyridine calcium antagonist with selective effects on the heart and peripheral circulation
  • Potentially viable neuroprotective agent for Parkinson's disease
Catalog No. 50-004-06124 Supplier Medchemexpress LLC Supplier No. HYB02335MG
Isradipine is an orally active, blood-brain barrier permeable L-type calcium channel blocker. It functions as a potent peripheral vasodilator and is a dihydropyridine calcium antagonist with selective effects on the heart and peripheral circulation. Isradipine is considered a potentially viable neuroprotective agent for Parkinson's disease.
  • Orally active
  • Blood-brain barrier permeable
  • L-type calcium channel blocker
  • Potent peripheral vasodilator
  • Dihydropyridine calcium antagonist with selective effects on the heart and peripheral circulation
  • Potentially viable neuroprotective agent for Parkinson's disease