Learn More
Medchemexpress LLC Isradipine | 75695-93-1 | 99.9% | 1 ML
SDP
Catalog No. 5000405720
Encompass_Preferred
Isradipine is an orally active and blood-brain barrier permeable L-type calcium channel blocker. It acts as a powerful peripheral vasodilator and is a dihydropyridine calcium antagonist with selective actions on both the heart and peripheral circulation. It is considered a potentially viable neuroprotective agent for Parkinson's disease.
  • Orally active and blood-brain barrier permeable
  • L-type calcium channel blocker
  • Powerful peripheral vasodilator
  • Dihydropyridine calcium antagonist
  • Selective actions on heart and peripheral circulation
  • Potentially neuroprotective agent for Parkinson's disease
  • Targets calcium channel and autophagy
  • Used in cardiovascular and neurological disease research
Catalog No. 50-004-05720 Supplier Medchemexpress LLC Supplier No. HYB023310MM/1ML
Isradipine is an orally active and blood-brain barrier permeable L-type calcium channel blocker. It acts as a powerful peripheral vasodilator and is a dihydropyridine calcium antagonist with selective actions on both the heart and peripheral circulation. It is considered a potentially viable neuroprotective agent for Parkinson's disease.
  • Orally active and blood-brain barrier permeable
  • L-type calcium channel blocker
  • Powerful peripheral vasodilator
  • Dihydropyridine calcium antagonist
  • Selective actions on heart and peripheral circulation
  • Potentially neuroprotective agent for Parkinson's disease
  • Targets calcium channel and autophagy
  • Used in cardiovascular and neurological disease research