Learn More
Apexbio Technology LLC Isradipine (Dynacirc)(Synonyms: Dynacirc, Prescal, Lomir, PN 200-110, Isrodipine, (+/-)-Isradipine, Isradipex), 10mM (in 1mL DMSO), CAS: 75695-93-1.
SDP
Catalog No. 5000060585
Encompass_Preferred
Isradipine (Dynacirc CAS 75695-93-1) is a small molecule antagonist belonging to the dihydropyridine class of calcium channel blockers It functions primarily by inhibiting L-type voltage-gated calcium channels thereby decreasing intracellular calcium influx into cardiac and vascular smooth muscle cells This inhibitory effect relaxes vascular smooth muscle tissue leading to vasodilation and subsequent reduction in systemic blood pressure In biomedical research isradipine is studied for its potential neuroprotective properties related to calcium-mediated excitotoxicity offering implications for conditions associated with neuronal calcium imbalance
Catalog No. 50-000-60585 Supplier Apexbio Technology LLC Supplier No. A84535.1
Isradipine (Dynacirc CAS 75695-93-1) is a small molecule antagonist belonging to the dihydropyridine class of calcium channel blockers It functions primarily by inhibiting L-type voltage-gated calcium channels thereby decreasing intracellular calcium influx into cardiac and vascular smooth muscle cells This inhibitory effect relaxes vascular smooth muscle tissue leading to vasodilation and subsequent reduction in systemic blood pressure In biomedical research isradipine is studied for its potential neuroprotective properties related to calcium-mediated excitotoxicity offering implications for conditions associated with neuronal calcium imbalance