Learn More
BPS Bioscience Inc ISRE Luciferase Reporter HEK293 Cell Line (JAK/STAT Signaling Pathway) Size: 2 vials
SDP
Catalog No. NC3920858
Encompass
The ISRE Luciferase Reporter HEK293 Cell Line (JAK/STAT Signaling Pathway) is a HEK293 cell line designed to monitor the activity of the JAK (Janus kinase)/STAT (Signal Transducer and Activator of Transcription) signaling pathway. These cells expresses a firefly luciferase reporter driven by ISRE (interferon stimulated response element) stably integrated into HEK293 cells.This cell line has been shown to be responsive to stimulation with IFNα (interferon Alpha) and to treatment with the JAK inhibitor Pyridone 6.
Catalog No. NC3920858 Supplier BPS Bioscience Inc Supplier No. 60510
The ISRE Luciferase Reporter HEK293 Cell Line (JAK/STAT Signaling Pathway) is a HEK293 cell line designed to monitor the activity of the JAK (Janus kinase)/STAT (Signal Transducer and Activator of Transcription) signaling pathway. These cells expresses a firefly luciferase reporter driven by ISRE (interferon stimulated response element) stably integrated into HEK293 cells.This cell line has been shown to be responsive to stimulation with IFNα (interferon Alpha) and to treatment with the JAK inhibitor Pyridone 6.