Learn More
Gentarget ISRE-RLUC LENTIVIRUS IN PBS
Catalog No. NC4036264
Encompass
Pre-made lentivirus express firefly Luciferase 3 reporter under a minimal promoter inserted with 8 tandem repeats of ISRE /Interferon Stimulated Response Element/ for detecting JAK-STAT signaling pathway acitvity.
Catalog No. NC4036264 Supplier Gentarget Supplier No. LVP940NPBS
Pre-made lentivirus express firefly Luciferase 3 reporter under a minimal promoter inserted with 8 tandem repeats of ISRE /Interferon Stimulated Response Element/ for detecting JAK-STAT signaling pathway acitvity.