Learn More
TARGETMOL CHEMICALS INC ISRIB-A15 5MG
SDP
Catalog No. 502579261
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. ISRIB-A15 (ISR-IN-2) is an potent inhibitor of the integrated stress response with an EC50 of 0.8 nM. purity: 98%
Catalog No. 50-257-9261 Supplier TARGETMOL CHEMICALS INC Supplier No. T677565MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. ISRIB-A15 (ISR-IN-2) is an potent inhibitor of the integrated stress response with an EC50 of 0.8 nM. purity: 98%