Learn More
Apexbio Technology LLC ISRIB (trans-isomer) 1597403-47-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063076
Encompass_Preferred
ISRIB (trans-isomer) (CAS 1597403-47-8) is a small molecule inhibitor of the integrated stress response (ISR) ISR activation involves the phosphorylation of eukaryotic initiation factor 2 (eIF2 ) by stress-sensitive kinases leading to reduced global protein synthesis and enhanced translation of select mRNAs ISRIB counteracts ISR signaling by reversing the translational suppression induced by phosphorylated eIF2 inhibiting production of ATF4 protein decreasing stress granule assembly and promoting polyribosome formation during endoplasmic reticulum (ER) stress ISRIB stabilizes and activates eIF2B by facilitating its dimerization Rodent studies demonstrate ISRIB s capacity to enhance hippocampal-dependent spatial and fear-related learning and memory highlighting its utility in neuroscience research
Catalog No. 50-000-63076 Supplier Apexbio Technology LLC Supplier No. B36995.1
ISRIB (trans-isomer) (CAS 1597403-47-8) is a small molecule inhibitor of the integrated stress response (ISR) ISR activation involves the phosphorylation of eukaryotic initiation factor 2 (eIF2 ) by stress-sensitive kinases leading to reduced global protein synthesis and enhanced translation of select mRNAs ISRIB counteracts ISR signaling by reversing the translational suppression induced by phosphorylated eIF2 inhibiting production of ATF4 protein decreasing stress granule assembly and promoting polyribosome formation during endoplasmic reticulum (ER) stress ISRIB stabilizes and activates eIF2B by facilitating its dimerization Rodent studies demonstrate ISRIB s capacity to enhance hippocampal-dependent spatial and fear-related learning and memory highlighting its utility in neuroscience research