Learn More
Medchemexpress LLC HY-12495 100mg , ISRIB (trans-isomer) CAS:1597403-47-8 Purity:98%
SDP
Catalog No. 501493435
Encompass_Preferred
Medchemexpress, HY-12495 100mg ISRIB (trans-isomer) CAS:1597403-47-8 Formula:C22H24Cl2N2O4 IC50: 5 nM (PERK) Purity:98% ISRIB (trans-isomer) is a potent inhibitor of PERK with an IC 50 of 5 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3435 Supplier Medchemexpress LLC Supplier No. HY12495100MG
Medchemexpress, HY-12495 100mg ISRIB (trans-isomer) CAS:1597403-47-8 Formula:C22H24Cl2N2O4 IC50: 5 nM (PERK) Purity:98% ISRIB (trans-isomer) is a potent inhibitor of PERK with an IC 50 of 5 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.