Learn More
Jeio Tech Inc Jeio Tech™ ISS-7200R, 198L Stackable Large Capacity Refrigerated Incubator Shaker (120V, 60Hz), PID Temperature Control, For High-Throughput Microbial Cultivation and Protein Expression
SDP
Catalog No. NC3358238
Encompass_Preferred
Jeio Tech ISS Refrigerating Incubator Shakers are high-performance, stackable units designed for laboratories that require both space efficiency and increased capacity. Ideal for cell culture, microbial incubation, and protein expression, the ISS series combines precise PID temperature control with smooth and uniform orbital shaking to support consistent sample growth. The refrigerated model offers an extended temperature control range, enabling both cooling and heating applications. The stackable design allows multiple units to be vertically arranged without compromising performance, making it an excellent solution for high-throughput workflows. With adjustable shaking speed, forced-air circulation, and a large viewing window, the ISS offers uniform incubation conditions, quiet operation, and user-friendly control for demanding applications in biotechnology, microbiology, and pharmaceutical research. Shaking platform required; sold separately.
Catalog No. NC3358238 Supplier Jeio Tech Inc Supplier No. AAH239215U
Add to Cart
Add to Cart
Request a Quote
Jeio Tech ISS Refrigerating Incubator Shakers are high-performance, stackable units designed for laboratories that require both space efficiency and increased capacity. Ideal for cell culture, microbial incubation, and protein expression, the ISS series combines precise PID temperature control with smooth and uniform orbital shaking to support consistent sample growth. The refrigerated model offers an extended temperature control range, enabling both cooling and heating applications. The stackable design allows multiple units to be vertically arranged without compromising performance, making it an excellent solution for high-throughput workflows. With adjustable shaking speed, forced-air circulation, and a large viewing window, the ISS offers uniform incubation conditions, quiet operation, and user-friendly control for demanding applications in biotechnology, microbiology, and pharmaceutical research. Shaking platform required; sold separately.