Learn More
PREOMICS INC iST-NHS 12x, 12 sample kit
Catalog No. NC1699277
Encompass
iST-NHS sample preparation is fully compatible with chemical labeling reagents. Allowing streamlined peptide processing, chemical labeling and an efficient peptide clean-up step. The iST-NHS method decreases hands-on time, sample loss and the amount of chemical label required.
Catalog No. NC1699277 Supplier PREOMICS INC Supplier No. P.O.00026
iST-NHS sample preparation is fully compatible with chemical labeling reagents. Allowing streamlined peptide processing, chemical labeling and an efficient peptide clean-up step. The iST-NHS method decreases hands-on time, sample loss and the amount of chemical label required.