Learn More
Medchemexpress LLC HY-15718A 5mg Medchemexpress, Istaroxime (hydrochloride) CAS:374559-48-5 Purity:>98%
SDP
Catalog No. 502029161
Encompass_Preferred
Medchemexpress, HY-15718A 5mg Istaroxime (hydrochloride) CAS:374559-48-5 Istaroxime hydrochloride is a Na+/K+-ATPase inhibitor (IC50=0.11 μM) and a sarcoplasmic/endoplasmic reticulum calcium ATPase 2 (SERCA 2) activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9161 Supplier Medchemexpress LLC Supplier No. HY15718A5MG
Medchemexpress, HY-15718A 5mg Istaroxime (hydrochloride) CAS:374559-48-5 Istaroxime hydrochloride is a Na+/K+-ATPase inhibitor (IC50=0.11 μM) and a sarcoplasmic/endoplasmic reticulum calcium ATPase 2 (SERCA 2) activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.