Learn More
Medchemexpress LLC HY-15718A 100mg , Istaroxime (hydrochloride) PST2744 (hydrochloride) CAS:374559-48-5 Purity:98%
SDP
Catalog No. 501493034
Encompass_Preferred
Medchemexpress, HY-15718A 100mg Istaroxime (hydrochloride) PST2744 (hydrochloride) CAS:374559-48-5 Formula:C21H33ClN2O3 IC50: 0.11 μM (Na + ,K + -ATPase) Purity:98% Istaroxime hydrochloride is a potent inhibitor of Na + ,K + -ATPase with IC 50 of 0.11 μM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3034 Supplier Medchemexpress LLC Supplier No. HY15718A100MG
Medchemexpress, HY-15718A 100mg Istaroxime (hydrochloride) PST2744 (hydrochloride) CAS:374559-48-5 Formula:C21H33ClN2O3 IC50: 0.11 μM (Na + ,K + -ATPase) Purity:98% Istaroxime hydrochloride is a potent inhibitor of Na + ,K + -ATPase with IC 50 of 0.11 μM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.