Learn More
AdipoGen Isthmin-1 human ELISA Kit
SDP
Catalog No. 502562728
Encompass_Preferred
Colorimetric Sandwich ELISA Assay. Detects human Isthmin-1 in plasma and cell culture supernatant. It detects also mouse Isthmin-1 very conserved. Range 0.625 to 40ng/ml. Sensitivity 0.4 ng/ml. Samples Cell Culture Supernatant, Plasma. Isthmin-1 ISM1 was first identified as a gene expressed in the Xenopus midbrain-hindbrain organizer called the isthmus, with a proposed role during early brain development. Isthmin-1 encodes a predicted ~50-kDa protein containing a signal peptide, a thrombospondin domain and an adhesion-associated domain. Isthmin-1 is important for embryonic and postnatal development. Growing evidence has shown that aberrant expression of Isthmin-1 can also affect the biological behavior of cancer. The Ism1 gene is conserved in mice and humans. A recent study showed that Isthmin-1 is an adipokine that induces glucose uptake in human and mouse adipocytes.
Catalog No. 50-256-2728 Supplier AdipoGen Supplier No. AG45B0032KI01
Add to Cart
Add to Cart
Colorimetric Sandwich ELISA Assay. Detects human Isthmin-1 in plasma and cell culture supernatant. It detects also mouse Isthmin-1 very conserved. Range 0.625 to 40ng/ml. Sensitivity 0.4 ng/ml. Samples Cell Culture Supernatant, Plasma. Isthmin-1 ISM1 was first identified as a gene expressed in the Xenopus midbrain-hindbrain organizer called the isthmus, with a proposed role during early brain development. Isthmin-1 encodes a predicted ~50-kDa protein containing a signal peptide, a thrombospondin domain and an adhesion-associated domain. Isthmin-1 is important for embryonic and postnatal development. Growing evidence has shown that aberrant expression of Isthmin-1 can also affect the biological behavior of cancer. The Ism1 gene is conserved in mice and humans. A recent study showed that Isthmin-1 is an adipokine that induces glucose uptake in human and mouse adipocytes.