Learn More
AdipoGen Isthmin-1 mAb Giusepi-1-4
SDP
Supplier:  AdipoGen AG27B0022C100
Encompass_Preferred
Recombinant Antibody. Recognizes human and mouse Isthmin-1. Isotype Mouse IgG2blambda. Immunogen Recombinant human Isthmin-1. Applications ELISA, WB. Liquid. In PBS and 0.02% Proclin-300. Isthmin-1 ISM1 was first identified as a gene expressed in the Xenopus midbrain hind brain organizer called isthmus, with a proposed role during early brain development. Isthmin-1 encodes a predicted ~50-kDa protein containing a signal peptide, a thrombospondin domain and an adhesion-associated domain. Isthmin-1 is important for embryonic and postnatal development. Growing evidence has shown that aberrant expression of Isthmin-1 can also affect the biological behavior of cancer. The Ism1 gene is conserved in mice and humans. A recent study showed that Isthmin-1 is an adipokine that induces glucose uptake in human and mouse adipocytes.
Catalog No. 50-256-3148