Learn More
Cayman Chemical Istradefylline-13C-d3, Purity: ≥99% deuterated forms (d1-d3), 1 mg
SDP
Catalog No. 502989987
Encompass_Preferred
An internal standard for the quantification of istradefylline by GC- or LC-MS
Catalog No. 50-298-9987 Supplier Cayman Chemical Supplier No. 312401 MG
An internal standard for the quantification of istradefylline by GC- or LC-MS